2-Nitrophenylisothiocyanat ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

L09669.06
L09669.06EA 109 EUR
L09669.06
2-Nitrophenylisothiocyanat ≥97%
2-Nitrophenylisothiocyanat

Purity: 97% CAS No.: 2719-30-4 Molecular Formula: C7H4N2O2S Formula Weight: 180.19 Flash Point: 110°(230°F) Odour: Irritating Boiling Point: 135-137°/17mm Beilstein Reference: 2719657 UN No.: 2923 MDL No.: MFCD00007092

Formel: C₇H₄N₂O₂S
Molecular Weight: 180.19 g/mol
Siedepunkt: 135…137 °C (17 mmHg)
Schmelzpunkt: 73…76 °C
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041238
CAS-Nummer: 2719-30-4
UN: 2923
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR