Wasser, Zellkultur-Qualität pyrogenfrei

Lieferant: Biowest

L0970-500 L0970-1000 L0970-100
L0970-500EA 8.4 EUR
L0970-500 L0970-1000 L0970-100
Wasser, Zellkultur-Qualität pyrogenfrei
Wasser

Cell culture water is high-quality water suitable for use as a solvent in the preparation of cell culture media and laboratory reagents.


  • Pyrogen-free

Formel: H₂O
Siedepunkt: 100 °C (1013 hPa)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³ (4 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR