Wasser, Zellkultur-Qualität pyrogenfrei
Lieferant: Biowest
L0970-500
L0970-1000
L0970-100
L0970-500EA
8.4
EUR
L0970-500
L0970-1000
L0970-100
Wasser, Zellkultur-Qualität pyrogenfrei
Wasser
Cell culture water is high-quality water suitable for use as a solvent in the preparation of cell culture media and laboratory reagents.
- Pyrogen-free
Formel:
H₂O Siedepunkt: 100 °C (1013 hPa) Schmelzpunkt: 0 °C Dichte: 1 g/cm³ (4 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00011332 CAS-Nummer: 7732-18-5 EINECS: 231-791-2 Merck Index: 13,10098 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...