Tetramethoxysilan ≥98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L09716.14
L09716.22
L09716.36
L09716.C4
L09716.14EA
26
EUR
L09716.14
L09716.22
L09716.36
L09716.C4
Tetramethoxysilan ≥98%
Tetramethoxysilan
Purity: 98%
CAS No.: 681-84-5
Molecular Formula: C4H12O4Si
Formula Weight: 152.22
Flash Point: 26°(78°F)
Refractive Index: 1.3680
Odour: Ether-like
Melting Point: 4-5°
Boiling Point: 121-122°
Beilstein Reference: 1699658
UN No.: 2606
MDL No.: MFCD00008341
Formel:
C₄H₁₂O₄Si Siedepunkt: 121…122 °C Schmelzpunkt: 4…5 °C Dichte: 1,032 Flammpunkt: 26 °C (78 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008341 CAS-Nummer: 681-84-5 EINECS: 211-656-4 UN: 2606 ADR: 6.1,I |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...