1,2-Dimethylnaphthalin ≥98%
Lieferant: Thermo Fisher Scientific
L09774.03
L09774.MD
L09774.03EA
76.8
EUR
L09774.03
L09774.MD
1,2-Dimethylnaphthalin ≥98%
1,2-Dimethylnaphthalene
Purity: 98%
CAS No.: 573-98-8
Molecular Formula: C12H12
Formula Weight: 156.23
Flash Point:
Formel:
C₁₂H₁₂ Siedepunkt: 266…267 °C Dichte: 1,013 Flammpunkt: >110 °C (230 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00004035 CAS-Nummer: 573-98-8 EINECS: 209-364-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...