(Aminomethyl)phosphonsäure 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
(Aminomethyl)phosphonic Acid, AMPA
L09833.03
L09833.MD
L09833.06
L09833.03EA
142
EUR
L09833.03
L09833.MD
L09833.06
(Aminomethyl)phosphonsäure 99%
(Aminomethyl)phosphonic acid
Purity: 99%
CAS No.: 1066-51-9
Molecular Formula: CH6NO3P
Formula Weight: 111.04
Melting Point: ca 290° dec.
Beilstein Reference: 1701219
Formel:
CH₆NO₃P Schmelzpunkt: ∼ 290 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00008105 CAS-Nummer: 1066-51-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...