(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L09834.09
L09834.04
L09834.09EA
114
EUR
L09834.09
L09834.04
(2S)-5,7-Dihydroxy-2-(4-hydroxyphenyl)-2,3-dihydro-4H-chromen-4-on ≥97%
(±)-Naringenin
Purity: 97%
CAS No.: 67604-48-2
Molecular Formula: C15H12O5
Formula Weight: 272.26
Melting Point: 248-251°
Merck Reference: 14,6424
MDL No.: MFCD00006844
Formel:
C₁₅H₁₂O₅ Schmelzpunkt: 248…251 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006844 CAS-Nummer: 67604-48-2 EINECS: 266-769-1 Merck Index: 14,6424 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...