4-Chlor-2-(trifluormethylphenyl)isocyanat

Lieferant: Thermo Fisher Scientific

L09850.03
L09850.03EA 29.8 EUR
L09850.03
4-Chlor-2-(trifluormethylphenyl)isocyanat
4-Chlor-2-(trifluormethylphenyl)isocyanat

Purity: 97%
CAS No.: 16588-69-5
Molecular Formula: C8H3ClF3NO
Formula Weight: 221.57
Storage Temperature: 2°C to 8°C
Flash Point: 94°(201°F)
Refractive Index: 1.5010
Odour: Pungent
Boiling Point: 114-115°/10mm
Beilstein Reference: 2214336
UN No.: 2206
MDL No.: MFCD00013873

Formel: C₈H₃ClF₃NO
Molecular Weight: 221.57 g/mol
Dichte: 1.4666 g/cm³ (20 °C)
MDL: MFCD00013873
CAS-Nummer: 16588-69-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR