4-Chlor-2-(trifluormethylphenyl)isocyanat 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
L09850.06
L09850.06EA
96.5
EUR
L09850.06
4-Chlor-2-(trifluormethylphenyl)isocyanat 97%
4-Chlor-2-(trifluormethylphenyl)isocyanat
Purity: 97%
CAS No.: 16588-69-5
Molecular Formula: C8H3ClF3NO
Formula Weight: 221.57
Storage Temperature: 2°C to 8°C
Flash Point: 94°(201°F)
Refractive Index: 1.5010
Odour: Pungent
Boiling Point: 114-115°/10mm
Beilstein Reference: 2214336
UN No.: 2206
MDL No.: MFCD00013873
Formel:
C₈H₃ClF₃NO Molecular Weight: 221.57 g/mol Siedepunkt: 114…115 °C (10 mmHg) Dichte: 1.467 Flammpunkt: 94 °C (201 °F) |
MDL:
MFCD00013873 CAS-Nummer: 16588-69-5 UN: 2206 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...