Dibenzylphosphonat 90+%

Lieferant: Thermo Fisher Scientific
Achtung

L09858.06
L09858.06EA 29.5 EUR
L09858.06
Dibenzylphosphonat 90+%
Dibenzylphosphonat

Purity: 90+% CAS No.: 17176-77-1 Molecular Formula: C14H15O3P Formula Weight: 262.25 Flash Point: 110°(230°F) Refractive Index: 1.5540 Odour: Odourless Boiling Point: 110-120°/0.1mm Beilstein Reference: 1982794 Merck Reference: 14,3014 MDL No.: MFCD00004774

Formel: C₁₄H₁₅O₃P
Siedepunkt: 110…120 °C (0,1 mmHg)
Dichte: 1,187
Flammpunkt: >110 °C (230 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00004774
CAS-Nummer: 17176-77-1
EINECS: 241-226-1
Merck Index: 14,3014

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR