4-Chlor-3-nitrophenol ≥99%

Lieferant: Thermo Fisher Scientific
Achtung

L10108.06 L10108.14
L10108.06EA 40.6 EUR
L10108.06 L10108.14
4-Chlor-3-nitrophenol ≥99%
4-Chlor-3-nitrophenol

Purity: 99% CAS No.: 610-78-6 Molecular Formula: C6H4ClNO3 Formula Weight: 173.56 Odour: Characteristic Melting Point: 124-129° Beilstein Reference: 2094546 MDL No.: MFCD00043546

Formel: C₆H₄ClNO₃
Schmelzpunkt: 124…129 °C
Dichte: 1 g/cm³ (20 °C)
Lagertemperatur: Raumtemperatur
MDL: MFCD00043546
CAS-Nummer: 610-78-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR