Phenyl trifluoromethyl sulphide 98%

Lieferant: Thermo Fisher Scientific
Achtung

L10114.03 L10114.06
L10114.03EA 30.2 EUR
L10114.03 L10114.06
Phenyl trifluoromethyl sulphide 98%
Phenyl trifluoromethyl sulphide

Purity: 98% CAS No.: 456-56-4 Molecular Formula: C7H5F3S Formula Weight: 178.18 Flash Point: 37°(98°F) Refractive Index: 1.4655 Odour: Unpleasant Boiling Point: 140-142° Beilstein Reference: 2043134 UN No.: UN1993 MDL No.: MFCD00040839

Formel: C₇H₅F₃S
Siedepunkt: 140…142 °C
Dichte: 1,249
Flammpunkt: 37 °C (98 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040839
CAS-Nummer: 456-56-4
EINECS: 207-270-0
ADR: 3,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR