4-(Trifluoracetyl)benzoesäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-(2,2,2-trifluoroacetyl)benzoic acid

L10242.03 L10242.MD
L10242.03EA 86.5 EUR
L10242.03 L10242.MD
4-(Trifluoracetyl)benzoesäure ≥97%
4-(Trifluoracetyl)benzoesäure

Purity: 97+% CAS No.: 58808-59-6 Molecular Formula: C9H5F3O3 Formula Weight: 218.13 Melting Point: 177-179° Beilstein Reference: 2728778

Formel: C₉H₅F₃O₃
Molecular Weight: 218.13 g/mol
Schmelzpunkt: 177…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00052340
CAS-Nummer: 58808-59-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR