cis-2,3-Epoxybutan ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
L10320.06
L10320.06EA
125
EUR
L10320.06
cis-2,3-Epoxybutan ≥98%
cis-2,3-Epoxybutane
Purity: 98%
CAS No.: 1758-33-4
Molecular Formula: C4H8O
Formula Weight: 72.11
Flash Point: -21°(-5°F)
Refractive Index: 1.3830
Melting Point: -84°
Boiling Point: 60-61°
Beilstein Reference: 79770
UN No.: 3271
MDL No.: MFCD00005128
Formel:
C₄H₈O Molecular Weight: 72,11 g/mol Siedepunkt: 60…61 °C Schmelzpunkt: –84 °C Dichte: 0,826 Flammpunkt: –21 °C (–5 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005128 CAS-Nummer: 1758-33-4 UN: 3271 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...