1,3-Cyclopentandion ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: Cyclopentane-1,3-dione

L10336.03 L10336.06
L10336.03EA 50.2 EUR
L10336.03 L10336.06
1,3-Cyclopentandion ≥98%
1,3-Cyclopentandion

Purity: 98% CAS No.: 3859-41-4 Molecular Formula: C5H6O2 Formula Weight: 98.10 Melting Point: 151-153° Beilstein Reference: 1362728

Formel: C₅H₆O₂
Molecular Weight: 98.1 g/mol
Schmelzpunkt: 151…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001405
CAS-Nummer: 3859-41-4
EINECS: 223-372-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR