(R)-(-)-1-Phenyl-1,2-ethandiol ≥99%

Lieferant: Thermo Scientific

Synonyme: (R)-(-)-Phenylethan-1,2-diol, (R)-(-)-α,β-Dihydroxyethylbenzol, (R)-(-)-Phenylethylenglycol, (R)-(-)-(1,2-Dihydroxyethyl)benzol

L10494.03
L10494.03EA 62.7 EUR
L10494.03
(R)-(-)-1-Phenyl-1,2-ethandiol ≥99%
(R)-(-)-1-Phenyl-1,2-ethandiol

Purity: 99% CAS No.: 16355-00-3 Molecular Formula: C8H10O2 Formula Weight: 138.17 Melting Point: 67-69° Boiling Point: 272-274° MDL No.: MFCD00064262

Formel: C₈H₁₀O₂
Molecular Weight: 138,17 g/mol
Siedepunkt: 272…274 °C
Schmelzpunkt: 67…69 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064262
CAS-Nummer: 16355-00-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR