1-(2-Hydroxyphenyl)-2-thioharnstoff ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L10519.03 L10519.06
L10519.03EA 19.4 EUR
L10519.03 L10519.06
1-(2-Hydroxyphenyl)-2-thioharnstoff ≥97%
1-(2-Hydroxyphenyl)-2-thioharnstoff

Purity: 97% CAS No.: 1520-26-9 Molecular Formula: C7H8N2OS Formula Weight: 168.22 Odour: Odourless Melting Point: ca 158° dec. Beilstein Reference: 1102479 MDL No.: MFCD00041159

Formel: (C₆H₄(OH))NC(S)NH₂
Siedepunkt: 323 °C (1013 hPa)
Schmelzpunkt: ∼ 158 °C (Zers.)
Lagertemperatur: Raumtemperatur
MDL: MFCD00041159
CAS-Nummer: 1520-26-9
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR