3-Methylquinoline-N-oxide 97%

Lieferant: Thermo Fisher Scientific

L10635.03
L10635.03EA 51.5 EUR
L10635.03
3-Methylquinoline-N-oxide 97%
3-Methylquinoline-N-oxide

Purity: 97% CAS No.: 1873-55-8 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 88-89° Beilstein Reference: 118125 MDL No.: MFCD00082650

Formel: C₁₀H₉NO
Schmelzpunkt: 88…89 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082650
CAS-Nummer: 1873-55-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR