5-(3-Chlorphenyl)-1H-tetrazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-(3-Chlorophenyl)-2H-1,2,3,4-tetrazole
L10699.03
L10699.03EA
39.6
EUR
L10699.03
5-(3-Chlorphenyl)-1H-tetrazol ≥98%
5-(3-Chlorphenyl)-1H-tetrazol
Purity: 98+%
CAS No.: 41421-28-7
Molecular Formula: C7H5ClN4
Formula Weight: 180.60
Melting Point: 139-140°
Beilstein Reference: 140081
Formel:
C₇H₅ClN₄ Molecular Weight: 180,6 g/mol Schmelzpunkt: 139…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00040951 CAS-Nummer: 41421-28-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...