2-Indanylacetic acid 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-(2,3-Dihydro-1H-inden-2-yl)acetic acid

L10700.03 L10700.06
L10700.03EA 96 EUR
L10700.03 L10700.06
2-Indanylacetic acid 99%
2-Indanylacetic acid

Purity: 99% CAS No.: 37868-26-1 Molecular Formula: C11H12O2 Formula Weight: 176.22 Melting Point: 91-93° Beilstein Reference: 2616409 MDL No.: MFCD00082653

Formel: C₁₁H₁₂O₂
Schmelzpunkt: 91…93 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00082653
CAS-Nummer: 37868-26-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR