2-Indanylacetic acid 99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
2-(2,3-Dihydro-1H-inden-2-yl)acetic acid
L10700.03
L10700.06
L10700.03EA
96
EUR
L10700.03
L10700.06
2-Indanylacetic acid 99%
2-Indanylacetic acid
Purity: 99%
CAS No.: 37868-26-1
Molecular Formula: C11H12O2
Formula Weight: 176.22
Melting Point: 91-93°
Beilstein Reference: 2616409
MDL No.: MFCD00082653
Formel:
C₁₁H₁₂O₂ Schmelzpunkt: 91…93 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082653 CAS-Nummer: 37868-26-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...