(±)-2,4-Difluormandelsäure ≥97%

Lieferant: Thermo Fisher Scientific
Achtung

L10708.03 L10708.06
L10708.03EA 63.8 EUR
L10708.03 L10708.06
(±)-2,4-Difluormandelsäure ≥97%
(±)-2,4-Difluormandelsäure

Purity: 97% CAS No.: 132741-30-1 Molecular Formula: C8H6F2O3 Formula Weight: 188.13 Melting Point: 122-124° MDL No.: MFCD00061296

Formel: C₈H₆F₂O₃
Schmelzpunkt: 122…124 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00061296
CAS-Nummer: 132741-30-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR