1,1-Dibrom-3,3,3-trifluoraceton ≥95%
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
3,3-Dibromo-1,1,1-trifluoroacetone, 3,3-Dibromo-1,1,1-trifluoro-2-propanone, 3,3-Dibromo-1,1,1-trifluoropropan-2-one
L10826.06
L10826.06EA
78.8
EUR
L10826.06
1,1-Dibrom-3,3,3-trifluoraceton ≥95%
1,1-Dibrom-3,3,3-trifluoraceton
Purity: 95%
CAS No.: 431-67-4
Molecular Formula: C3HBr2F3O
Formula Weight: 269.85
Storage Temperature: 2°C to 8°C
Refractive Index: 1.4305
Odour: Pungent
Boiling Point: 111-113°
Beilstein Reference: 636645
UN No.: 3265
MDL No.: MFCD00041362
Formel:
Br₂HCC(O)CF₃ Molecular Weight: 269,84 g/mol Siedepunkt: 111…113 °C Dichte: 2,188 g/cm³ (20 °C) Lagertemperatur: Kühlschrank |
MDL:
MFCD00041362 CAS-Nummer: 431-67-4 UN: 3265 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...