Amylacetat ≥99%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Essigsäureamylester, Essigsäurepentylester, Pentylacetat
L10852.22
L10852.36
L10852.22EA
25.2
EUR
L10852.22
L10852.36
Amylacetat ≥99%
Amylacetat
Purity: 99%
CAS No.: 628-63-7
Molecular Formula: CH3CO2(CH2)4CH3
Formula Weight: 130.19
Flash Point: 24°(75°F)
Refractive Index: 1.4030
Odour: Fruit-like
Melting Point: -100°
Boiling Point: 142°
Beilstein Reference: 1744753
UN No.: 1104
MDL No.: MFCD00009500
Formel:
C₇H₁₄O₂ Molecular Weight: 130.19 g/mol Siedepunkt: 142 °C Schmelzpunkt: –100 °C Dichte: 0.876 Flammpunkt: 24 °C (75 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00009500 CAS-Nummer: 628-63-7 EINECS: 211-047-3 UN: 1104 ADR: 3 III |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...