1,1,1-Trifluor-2-methyl-2-propanol 98%

Lieferant: Thermo Scientific
Gefahr

Synonyme: 1,1,1-Trifluoro-2-methylpropan-2-ol

L10888.06
L10888.06EA 80.4 EUR
L10888.06
1,1,1-Trifluor-2-methyl-2-propanol 98%
2-Trifluoromethyl-2-propanol

Purity: 98% CAS No.: 507-52-8 Molecular Formula: C4H7F3O Formula Weight: 128.09 Flash Point: 15°(59°F) Refractive Index: 1.3400 Odour: Recognizable Boiling Point: 80-81° Beilstein Reference: 1736196 UN No.: UN1987 MDL No.: MFCD00014391

Formel: C₄H₇F₃O
Molecular Weight: 128,09 g/mol
Siedepunkt: 80…81 °C
Dichte: 1,170
Flammpunkt: 15 °C (59 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00014391
CAS-Nummer: 507-52-8
EINECS: 208-075-3
ADR: 3,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR