Methyl-4-(methylthio)butyrate 96%

Lieferant: Thermo Scientific
Achtung

L11000.06
L11000.06EA 56.4 EUR
L11000.06
Methyl-4-(methylthio)butyrate 96%
Methyl-4-(methylthio)butyrate

Purity: 96% CAS No.: 53053-51-3 Molecular Formula: CH3S(CH2)3CO2CH3 Formula Weight: 148.23 Flash Point: 72°(161°F) Refractive Index: 1.4660 Boiling Point: 114-115°/40mm MDL No.: MFCD00009849

Formel: CH₃S(CH₂)₃CO₂CH₃
Molecular Weight: 148,23 g/mol
Siedepunkt: 114…115 °C (40 mmHg)
Dichte: 1,036
Flammpunkt: 72 °C (161 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00009849
CAS-Nummer: 53053-51-3
EINECS: 258-328-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR