(R)-(-)-2-Amino-1-propanol 98%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
D(-)-Alaninol, D(-)-2-Amino-1-propanol
L11030.03
L11030.06
L11030.14
L11030.03EA
41.7
EUR
L11030.03
L11030.06
L11030.14
(R)-(-)-2-Amino-1-propanol 98%
(R)-(-)-2-Amino-1-propanol
Purity: 98%
CAS No.: 35320-23-1
Molecular Formula: C3H9NO
Formula Weight: 75.11
Flash Point: 83°(181°F)
Refractive Index: 1.4493
Boiling Point: 174-176°
Beilstein Reference: 1718866
Optical Rotation: -22° (c=2 in ethanol)
UN No.: 2735
MDL No.: MFCD00064413
Formel:
C₃H₉NO Molecular Weight: 75.11 g/mol Siedepunkt: 174…176 °C Dichte: 0.965 Flammpunkt: 83 °C (181 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00064413 CAS-Nummer: 35320-23-1 ADR: 8 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...