2-Amino-5-methylthiazol ≥98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5-Methylthiazol-2-ylamin
L11039.06
L11039.06EA
22.5
EUR
L11039.06
2-Amino-5-methylthiazol ≥98%
2-Amino-5-methylthiazole
Purity: 98+%
CAS No.: 7305-71-7
Molecular Formula: C4H6N2S
Formula Weight: 114.17
Odour: Recognizable
Melting Point: 95-98°
Beilstein Reference: 109603
UN No.: 3077
MDL No.: MFCD00078317
Formel:
C₄H₆N₂S Molecular Weight: 114,17 g/mol Siedepunkt: 222 °C (1013 hPa) Schmelzpunkt: 95…98 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00078317 CAS-Nummer: 7305-71-7 EINECS: 423-800-5 ADR: 9,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...