2,4-Dichlorphenylisocyanat 96%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2,4-Dichlorophenylisocyantae

L11089.06 L11089.14 L11089.03
L11089.06EA 52.1 EUR
L11089.06 L11089.14 L11089.03
2,4-Dichlorphenylisocyanat 96%
2,4-Dichlorphenylisocyanat

Purity: 96% CAS No.: 2612-57-9 Molecular Formula: C7H3Cl2NO Formula Weight: 188.01 Odour: Irritating Melting Point: 57-61° Boiling Point: 80°/1mm Beilstein Reference: 2090780 UN No.: 2250 MDL No.: MFCD00002001

Formel: C₇H₃Cl₂NO
Siedepunkt: 80 °C (1 mmHg)
Schmelzpunkt: 57…61 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00002001
CAS-Nummer: 2612-57-9
EINECS: 220-040-4
UN: 2250
ADR: 6.1,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR