Ethyl-4-(trifluormethyl)benzoat ≥99%
Lieferant: Thermo Fisher Scientific
L11139.03
L11139.03EA
22.6
EUR
L11139.03
Ethyl-4-(trifluormethyl)benzoat ≥99%
Ethyl-4-(trifluormethyl)benzoat
Purity: 99%
CAS No.: 583-02-8
Molecular Formula: C10H9F3O2
Formula Weight: 218.18
Refractive Index: 1.4470
Boiling Point: 78-80°/5mm
MDL No.: MFCD00013563
Formel:
C₁₀H₉F₃O₂ Siedepunkt: 78…80 °C (5 mmHg) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00013563 CAS-Nummer: 583-02-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...