5-Bromoindoxyl diacetate ≥98%

Lieferant: Thermo Fisher Scientific

Synonyme: 5-Bromoindolyl-1,3-diacetate, 1-Acetyl-5-bromo-1H-indol-3-yl acetate

L11189.03
L11189.03EA 59.5 EUR
L11189.03
5-Bromoindoxyl diacetate ≥98%
5-Bromoindoxyl diacetate

Purity: 98+% CAS No.: 33588-54-4 Molecular Formula: C12H10BrNO3 Formula Weight: 296.13 Storage Temperature: 2°C to 8°C Melting Point: 122-125° Beilstein Reference: 236935 MDL No.: MFCD00005799

Formel: C₁₂H₁₀BrNO₃
Schmelzpunkt: 122…125 °C
MDL: MFCD00005799
CAS-Nummer: 33588-54-4
EINECS: 251-584-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR