Pyranine

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: HPTS, 8-Hydroxy-1,3,6-pyrenetrisulphonic acid trisodium salt, Japan Green No. 204, Sodium 8-hydroxypyrene-1,3,6-trisulphonate, Trisodium 8-hydroxypyrene-1,3,6-trisulphonate

L11252.06 L11252.14
L11252.06EA 43.8 EUR
L11252.06 L11252.14
Pyranine
Pyranine

CAS No.: 6358-69-6 Molecular Formula: C16H7Na3O10S3 Formula Weight: 524.39 Beilstein Reference: 4107272 MDL No.: MFCD00037575

Formel: C₁₆H₇Na₃O₁₀S₃
Molecular Weight: 524.371 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037575
CAS-Nummer: 6358-69-6
EINECS: 228-783-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR