γ-Cyclodextrin Hydrat
Lieferant: Thermo Fisher Scientific
CAS No.: 91464-90-3
Molecular Formula: C48H80O40.xH2O
Formula Weight: 1297.15(anhy)
Melting Point: ca 267° dec.
Beilstein Reference: 5725162
Merck Reference: 14,2718
Optical Rotation: +177° (c=1 in water) on dry basis
MDL No.: MFCD00149574
Formel:
C₄₈H₈₀O₄₀·H₂O Lagertemperatur: Raumtemperatur |
MDL:
MFCD00149574 CAS-Nummer: 91464-90-3 EINECS: 241-482-4 Merck Index: 14,2718 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...