trans-1,2-Cyclohexandicarbonsäure ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: trans-Cyclohexane-1,2-dicarboxylic acid

L11288.06 L11288.14
L11288.06EA 51.8 EUR
L11288.06 L11288.14
trans-1,2-Cyclohexandicarbonsäure ≥98%
trans-1,2-Cyclohexanedicarboxylic acid

Purity: 98% CAS No.: 2305-32-0 Molecular Formula: C8H12O4 Formula Weight: 172.18 Melting Point: 220-221° Beilstein Reference: 3200609

Formel: C₈H₁₂O₄
Siedepunkt: 339 °C (1013 hPa)
Schmelzpunkt: 220…221 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001464
CAS-Nummer: 2305-32-0
EINECS: 218-975-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR