4-Chloro-5-nitrophthalimide 95%
Lieferant: Thermo Fisher Scientific
L11308.03
L11308.03EA
105
EUR
L11308.03
4-Chloro-5-nitrophthalimide 95%
4-Chloro-5-nitrophthalimide
Purity: 95%
CAS No.: 6015-57-2
Molecular Formula: C8H3ClN2O4
Formula Weight: 226.58
Melting Point: 198-203°
Beilstein Reference: 1536189
MDL No.: MFCD00052331
Formel:
C₈H₃ClN₂O₄ Schmelzpunkt: 198…203 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052331 CAS-Nummer: 6015-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...