Chromotrop 2B ≥97%

Lieferant: Thermo Fisher Scientific

L11330.14 L11330.22
L11330.14EA 56 EUR
L11330.14 L11330.22
Chromotrop 2B ≥97%

CAS No.: 548-80-1 Molecular Formula: C16H9N3Na2O10S2 Formula Weight: 513.37 Melting Point: 300° Beilstein Reference: 4109733 Merck Reference: 14,2240 MDL No.: MFCD00003951

Formel: C₁₆H₉N₃Na₂O₁₀S₂
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003951
CAS-Nummer: 548-80-1
EINECS: 208-959-9
Merck Index: 14,2240

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR