Benzo[b]thiophene-2-carbohydrazide 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
L11357.03
L11357.06
L11357.03EA
64.8
EUR
L11357.03
L11357.06
Benzo[b]thiophene-2-carbohydrazide 97%
Benzo[b]thiophene-2-carbohydrazide
Purity: 97%
CAS No.: 175135-07-6
Molecular Formula: C9H8N2OS
Formula Weight: 192.24
Melting Point: 184-187°
Beilstein Reference: 153297
MDL No.: MFCD00052501
Formel:
C₉H₈N₂OS Schmelzpunkt: 184…187 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00052501 CAS-Nummer: 175135-07-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...