2-Amino-4-methoxybenzothiazol 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
4-Methoxybenzothiazol-2-ylamine
L11358.03
L11358.06
L11358.14
L11358.03EA
40.4
EUR
L11358.03
L11358.06
L11358.14
2-Amino-4-methoxybenzothiazol 97%
2-Amino-4-methoxybenzothiazole
Purity: 97%
CAS No.: 5464-79-9
Molecular Formula: C8H8N2OS
Formula Weight: 180.23
Melting Point: 153-159°
Beilstein Reference: 141363
Formel:
C₈H₈N₂OS Molecular Weight: 180.23 g/mol Siedepunkt: 335 Schmelzpunkt: 153…159 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005792 CAS-Nummer: 5464-79-9 EINECS: 226-763-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...