2-Brom-3-methylthiophen ≥97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Purity: 97%
CAS No.: 14282-76-9
Molecular Formula: C5H5BrS
Formula Weight: 177.07
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5710
Boiling Point: 173-176°
Beilstein Reference: 105719
UN No.: 2810
MDL No.: MFCD00059741
Formel:
C₅H₅BrS Siedepunkt: 173…176 °C Dichte: 1,580 Flammpunkt: 68 °C (154 °F) Lagertemperatur: Kühlschrank |
MDL:
MFCD00059741 CAS-Nummer: 14282-76-9 EINECS: 238-175-2 UN: 2810 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...