(3,5-Dimethoxyphenyl)acetonitril ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3,5-Dimethoxybenzylcyanid

L11443.03 L11443.06
L11443.03EA 57.8 EUR
L11443.03 L11443.06
(3,5-Dimethoxyphenyl)acetonitril ≥98%
(3,5-Dimethoxyphenyl)acetonitril

Purity: 98+% CAS No.: 13388-75-5 Molecular Formula: C10H11NO2 Formula Weight: 177.20 Odour: Characteristic Melting Point: 53-56° Beilstein Reference: 2096481 MDL No.: MFCD00016395

Formel: C₁₀H₁₁NO₂
Molecular Weight: 177.2
Siedepunkt: 298
Schmelzpunkt: 53…56 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00016395
CAS-Nummer: 13388-75-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR