2,3-Dichlorpyrazin ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

L11500.06
L11500.06EA 104 EUR
L11500.06
2,3-Dichlorpyrazin ≥98%
2,3-Dichloropyrazine

Purity: 98% CAS No.: 4858-85-9 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Refractive Index: 1.5645 Boiling Point: 105-107°/50mm MDL No.: MFCD00040964

Formel: C₄H₂Cl₂N₂
Siedepunkt: 105…107 °C (50 mmHg)
Lagertemperatur: Raumtemperatur
MDL: MFCD00040964
CAS-Nummer: 4858-85-9
EINECS: 225-460-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR