(±)-Allyl-2,3-epoxypropylether ≥97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Allyl glycidyl ether

L11634.22 L11634.36
L11634.22EA 33.6 EUR
L11634.22 L11634.36
(±)-Allyl-2,3-epoxypropylether ≥97%
(±)-Allyl-2,3-epoxypropylether

Purity: 97% CAS No.: 106-92-3 Molecular Formula: C6H10O2 Formula Weight: 114.15 Flash Point: 47°(134°F) Refractive Index: 1.4350 Odour: Ether-like Melting Point: -100° Boiling Point: 50-51°/15mm Beilstein Reference: 105871 UN No.: 2219 MDL No.: MFCD00005143

Formel: C₆H₁₀O₂
Molecular Weight: 114.14 g/mol
Siedepunkt: 50…51 °C (15 mmHg)
Schmelzpunkt: –100 °C
Dichte: 0.967
Flammpunkt: 47 °C (134 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00005143
CAS-Nummer: 106-92-3
EINECS: 203-442-4
UN: 2219
ADR: 3 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR