Isobornylacetat ≥94%
Lieferant: Thermo Fisher Scientific
Achtung
|
L11748.AE
L11748.AP
L11748.AEEA
28.2
EUR
L11748.AE
L11748.AP
Isobornylacetat ≥94%
Isobornylacetat
Purity: 94%
CAS No.: 125-12-2
Molecular Formula: C12H20O2
Formula Weight: 196.29
Flash Point: 89°(192°F)
Refractive Index: 1.4650
Boiling Point: 220-224°
Beilstein Reference: 3197572
MDL No.: MFCD00135943
Formel:
C₁₂H₂₀O₂ Siedepunkt: 220…224 °C Dichte: 0,990 Flammpunkt: 89 °C (192 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00135943 CAS-Nummer: 125-12-2 EINECS: 204-727-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...