Phenyl acrylate 97%

Lieferant: Thermo Fisher Scientific
Achtung

L11819.06 L11819.14
L11819.06EA 76.9 EUR
L11819.06 L11819.14
Phenyl acrylate 97%
Phenyl acrylate

Purity: 97% CAS No.: 937-41-7 Molecular Formula: C6H5O2CCH=CH2 Formula Weight: 148.16 Refractive Index: 1.5210 Boiling Point: 89-90°/12mm Beilstein Reference: 2041124 UN No.: UN3082 MDL No.: MFCD00048145

Formel: C₉H₈O₂
Siedepunkt: 89…90 °C (12 mmHg)
Dichte: 1,076
Lagertemperatur: Raumtemperatur
MDL: MFCD00048145
CAS-Nummer: 937-41-7
EINECS: 213-329-1
ADR: 9,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR