(S)-(-)-5-Hydroxymethyl-2(5H)-furanone 98%

Lieferant: Thermo Scientific

L11919.MD
L11919.MDEA 64.9 EUR
L11919.MD
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone 98%
(S)-(-)-5-Hydroxymethyl-2(5H)-furanone

Purity: 98% CAS No.: 78508-96-0 Molecular Formula: C5H6O3 Formula Weight: 114.10 Melting Point: 34-43° Beilstein Reference: 3537455 Optical Rotation: -148° (c=1.4 in water) MDL No.: MFCD00075370

Formel: C₅H₆O₃
Molecular Weight: 114,10 g/mol
Schmelzpunkt: 34…43 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075370
CAS-Nummer: 78508-96-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR