3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid ≥98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Thiazolyl blue tetrazolium bromide, MTT, 2-(4,5-Dimethylthiazol-2-yl)-3,5-diphenyl-2H-tetrazol-3-ium bromide

L11939.03 L11939.06
L11939.03EA 49.6 EUR
L11939.03 L11939.06
3-(4,5-Dimethyl-2-thiazolyl)-2,5-diphenyltetrazoliumbromid ≥98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

Purity: 98% CAS No.: 298-93-1 Molecular Formula: C18H16BrN5S Formula Weight: 414.33 Odour: Odourless Melting Point: ca 195° dec. Beilstein Reference: 3825277 MDL No.: MFCD00011964

Formel: C₁₈H₁₆N₅BrS
Schmelzpunkt: ∼ 195 °C (Zersetzung)
Lagertemperatur: Raumtemperatur
MDL: MFCD00011964
CAS-Nummer: 298-93-1
EINECS: 206-069-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR