4-Nitrophenyl octanoate 96%

Lieferant: Thermo Fisher Scientific

L12022.03 L12022.06
L12022.03EA 28.9 EUR
L12022.03 L12022.06
4-Nitrophenyl octanoate 96%
4-Nitrophenyl octanoate

Refractive Index: 1.5130

Formel: C₁₄H₁₉NO₄
Dichte: 1,082
Lagertemperatur: Raumtemperatur
MDL: MFCD00024665
CAS-Nummer: 1956-10-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR