5-Hydroxy-2-nitrobenzaldehyd ≥97%

Lieferant: Thermo Scientific
Achtung

L12051.06
L12051.06EA 89.8 EUR
L12051.06
5-Hydroxy-2-nitrobenzaldehyd ≥97%
5-Hydroxy-2-nitrobenzaldehyd

Purity: 97% CAS No.: 42454-06-8 Molecular Formula: C7H5NO4 Formula Weight: 167.12 Melting Point: 166-168° Beilstein Reference: 1910196

Formel: C₇H₅NO₄
Molecular Weight: 167,12 g/mol
Schmelzpunkt: 166…168 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00007332
CAS-Nummer: 42454-06-8
EINECS: 255-832-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR