1-(4-Methoxyphenyl)ethanol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 4-Methoxy-alpha-methylbenzyl alcohol

L12062.14 L12062.22
L12062.14EA 54.2 EUR
L12062.14 L12062.22
1-(4-Methoxyphenyl)ethanol 95%
1-(4-Methoxyphenyl)ethanol

Purity: 95% CAS No.: 3319-15-1 Molecular Formula: C9H12O2 Formula Weight: 152.19 Flash Point: 74°(165°F) Refractive Index: 1.5330 Boiling Point: 94-96°/1mm Beilstein Reference: 2043521 MDL No.: MFCD00016857

Formel: C₉H₁₂O₂
Siedepunkt: 94…96 °C (1 mmHg)
Dichte: 1,079
Flammpunkt: 74 °C (165 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00016857
CAS-Nummer: 3319-15-1
EINECS: 222-019-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR