Acetaldehyde ammonia trimer 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Aldehyde Ammonia Trimer, 2,4,6-Trimethylhexahydro-1,3,5-triazine Trihydrate, Hexahydro-2,4,6-trimethyl-1,3,5-triazine Trihydrate, 2,4,6-trimethyl-1,3,5-triazinane trihydrate

L12265.18
L12265.18EA 45.8 EUR
L12265.18
Acetaldehyde ammonia trimer 98%
Acetaldehyde ammonia trimer

Purity: 98% CAS No.: 58052-80-5 Molecular Formula: C6H15N3.3H2O Formula Weight: 183.25 Melting Point: 93-96° Boiling Point: 110° dec. Beilstein Reference: 3910586 UN No.: UN1841 MDL No.: MFCD00149559

Formel: C₆H₂₁N₃O₃
Molecular Weight: 183.25 g/mol
Siedepunkt: 110 °C (Zers.)
Schmelzpunkt: 93…96 °C
Dichte: 0.852
Lagertemperatur: Raumtemperatur
MDL: MFCD00149559
CAS-Nummer: 58052-80-5
EINECS: 200-868-2
ADR: 9 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR